Learn More
New England Biolabs, Inc. PvuII – 25000 units
SDP
Catalog No. 50812038
Encompass
A restriction endonuclease that recognizes the sequence CAG_^CTG.
Catalog No. 50-812-038 Supplier New England Biolabs, Inc. Supplier No. R0151M
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
A restriction endonuclease that recognizes the sequence CAG_^CTG.