Learn More
New England Biolabs, Inc. PvuII – 5000 units
SDP
Catalog No. 50812040
Encompass
A restriction endonuclease that recognizes the sequence CAG_^CTG.
Catalog No. 50-812-040 Supplier New England Biolabs, Inc. Supplier No. R0151T
A restriction endonuclease that recognizes the sequence CAG_^CTG.