Learn More
MSC P&G Professional Sanitizer and Cleaner, Powder, 10 lb, Chlorinated
Catalog No. 502839938
Encompass
This P&G Professional Clean Quick Chlorinated Sanitizer and Cleaner is a powder concentrate designed for effective cleaning and sanitizing. It is ideal for use with milkshake and soft-serve machines, as well as various flat foodservice surfaces. Its high phosphate formula actively helps prevent milk stone, ensuring thorough cleanliness and maintenance.
  • Designed for cleaning and sanitizing milkshake and soft-serve machines
  • High phosphate formula helps prevent milk stone
  • Provides 100 ppm chlorine when mixed at 1 oz. per 2 gallons of water
  • Powder concentrate for efficient use
  • Suitable for flat foodservice surfaces
  • EPA registered
  • Unscented formula
Catalog No. 50-283-9938 Supplier MSC Supplier No. 91482703
This P&G Professional Clean Quick Chlorinated Sanitizer and Cleaner is a powder concentrate designed for effective cleaning and sanitizing. It is ideal for use with milkshake and soft-serve machines, as well as various flat foodservice surfaces. Its high phosphate formula actively helps prevent milk stone, ensuring thorough cleanliness and maintenance.
  • Designed for cleaning and sanitizing milkshake and soft-serve machines
  • High phosphate formula helps prevent milk stone
  • Provides 100 ppm chlorine when mixed at 1 oz. per 2 gallons of water
  • Powder concentrate for efficient use
  • Suitable for flat foodservice surfaces
  • EPA registered
  • Unscented formula