Learn More
Aviva Systems Biology Corp PWP2 Antibody - middle region (ARP45822_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45822_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PWP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PWP2. Alternate names: EHOC-17, PWP2H, UTP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-59146