Learn More
Hach Company Power Supply, 115 Vac, 60 Hz, for Portable Ultraviolet Lamps 26708 & 26710
Catalog No. 501996512 Shop All Hach Company Products
Encompass
Shortwave (254 nm) UV lamps for field or laboratory use. Either lamp can be used inthe lab with power supply while Portable Field Pack can only be used with the1.75" lamp. Portable Field Pack requires a 9-Volt battery (not included) and has abelt clip for easy traveling. Both lamps are physically identical except for bulb and cord length (these are shorter in the 1.75" version). NOTE: Not for use in microbiological applications. UV Lamp contains mercury. Dispose according to state, local or federal regulations.
Catalog No. 50-199-6512 Supplier Hach Company Supplier No. 2670700
Shortwave (254 nm) UV lamps for field or laboratory use. Either lamp can be used inthe lab with power supply while Portable Field Pack can only be used with the1.75" lamp. Portable Field Pack requires a 9-Volt battery (not included) and has abelt clip for easy traveling. Both lamps are physically identical except for bulb and cord length (these are shorter in the 1.75" version). NOTE: Not for use in microbiological applications. UV Lamp contains mercury. Dispose according to state, local or federal regulations.