Learn More
Aviva Systems Biology Corp PWWP2A Antibody - C-terminal region (ARP48981_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48981_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PWWP2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PWWP2A. Alternate names: KIAA1935, MGC132770, MST101ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59983