Learn More
Selleck Chemical LLC PX-12 10mg 141400-58-0
SDP
Supplier:  Selleck Chemical LLC S794710MG
Encompass_Preferred
PX-12 is a potent thioredoxin-1 (Trx-1) inhibitor by irreversibly thioalkylation of Cys73 of Trx-1. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5461