Learn More
Medchemexpress LLC HY-13734 5mg Medchemexpress, PX-12 CAS:141400-58-0 Purity:>98%
SDP
Catalog No. 501969887
Encompass_Preferred
Medchemexpress, HY-13734 5mg PX-12 CAS:141400-58-0 PX-12(IV-2) is an irreversible inhibitor of Thioredoxin-1 (Trx-1); inhibits the growth of MCF-7 and HT-29 cells with IC50 values of 1.9 and 2.9 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9887 Supplier Medchemexpress LLC Supplier No. HY137345MG
Medchemexpress, HY-13734 5mg PX-12 CAS:141400-58-0 PX-12(IV-2) is an irreversible inhibitor of Thioredoxin-1 (Trx-1); inhibits the growth of MCF-7 and HT-29 cells with IC50 values of 1.9 and 2.9 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.