Learn More
Selleck Chemical LLC PX-478 2HCl
SDP
Catalog No. 5000004740
Encompass_Preferred
PX-478 2HCl is an orally active and selective hypoxia-inducible factor-1 (HIF-1 ) inhibitor PX-478 2HCl induces apoptosis and has anti-tumor activity Phase 1
Catalog No. 50-000-04740 Supplier Selleck Chemical LLC Supplier No. S76121MG5
PX-478 2HCl is an orally active and selective hypoxia-inducible factor-1 (HIF-1 ) inhibitor PX-478 2HCl induces apoptosis and has anti-tumor activity Phase 1