Learn More
Medchemexpress LLC HY-10231 50mg , PX-478 CAS:685898-44-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1023150MG
Encompass_Preferred
HY-10231 50mg PX-478 CAS: 685898-44-6 PX-478 is an orally active HIF-1α inhibitor with potent antitumor activities. PX-478 can cross the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9297