Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPB™ PX5™ PAPR Fan Housing
SDP

Catalog No. 17000147
Encompass_Preferred
Change view
Click to view available options
Product Type: Fan Housing; For Use With (Equipment): PX5™ PAPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-000-147 Product Type: Fan Housing; For Use With (Equipment): PX5™ PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING This product can expose you to chemicals including styrene, which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
Catalog No. 17-000-147 Supplier RPB™ Supplier No. 03810
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING This product can expose you to chemicals including styrene, which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.

Fan housing is a replacement part for the PX5™ PAPR.

  • Replacement part for the PX5™ PAPR

Specifications

Color Gray
Disposable Reusable
Material Plastic
Product Type Fan Housing
Size Small
For Use With (Equipment) PX5™ PAPR
Model 03-810
Dimensions (L x W x H) 8.7 x 6.8 x 2.5 in.
Weight (English) 1.04 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.