Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPB™ PX5™ PAPR Spark Arrestor
SDP

Catalog No. 17000158
Encompass_Preferred
Change view
Click to view available options
Product Type: Respirator Accessory; For Use With (Equipment): All PX5™ PAPR filters
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-000-158 Product Type: Respirator Accessory; For Use With (Equipment): All PX5™ PAPR filters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 17-000-158 Supplier RPB™ Supplier No. 03891

missing translation for 'validMainframeMsg'

WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

Spark arrestor can be used with all PX5™ respirator filters.

  • Replacement part for all PX5™ PAPR filters

Specifications

Color Moth Gray
Disposable Reusable
Material Stainless Steel
Product Type Spark Arrestor
Size One Size
For Use With (Equipment) All PX5™ PAPR Filters
Model 03-891
Dimensions (L x W x H) 7.4 x 3.9 x 0.1 in.
Weight (English) 0.02 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.