Learn More
ABclonal Technology PXDN Rabbit mAb
SDP
Catalog No. 5000008561
Encompass_Preferred
This gene encodes a heme-containing peroxidase that is secreted into the extracellular matrix It is involved in extracellular matrix formation and may function in the physiological and pathological fibrogenic response in fibrotic kidney Mutations in this gene cause corneal opacification and other ocular anomalies and also microphthalmia and anterior segment dysgenesis
Catalog No. 50-000-08561 Supplier ABclonal Technology Supplier No. A2677720UL
This gene encodes a heme-containing peroxidase that is secreted into the extracellular matrix It is involved in extracellular matrix formation and may function in the physiological and pathological fibrogenic response in fibrotic kidney Mutations in this gene cause corneal opacification and other ocular anomalies and also microphthalmia and anterior segment dysgenesis