Learn More
Aviva Systems Biology Corp Pxmp3 Antibody - N-terminal region (ARP42946_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42946_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pxmp3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 35kDa, D3Ertd138e, PMP35, Pex2, Pxmp3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-046