Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PXR (SXR) Ligand Binding Domain Protein, GST Tag Recombinant Protein

Catalog No. PV4841
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4841 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4841 Supplier Invitrogen™ Supplier No. PV4841
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Pregnane X Receptor (Steroid and Xenobiotic Receptor), Ligand Binding Domain (PXR (SXR)-LBD) is a 64.7 kDa recombinant human protein (amino acids 111-434) expressed as a GST fusion protein in baculovirus-infected insect cells. PXR (SXR)-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator recruitment or displacement studies. Protein Form: Recombinant, Ligand Binding Domain.

This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75469
Concentration See Label
For Use With (Application) Binding Assay
Formulation Proprietary buffer with no preservative
Gene ID (Entrez) 8856
Name PXR (SXR) Ligand Binding Domain Protein, GST Tag
Purification Method Purified
Quantity 50 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias BXR; mPXR; Nr1i2; Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group 1, member 2; nuclear receptor subfamily 1, group I, member 2; ONR 1; ONR1; orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; PAR; PAR 1; PAR 2; PAR q; PAR1; PAR2; PARq; pregnane X nuclear receptor variant 2; Pregnane X receptor; pregnane X receptor (nuclear receptor sub family 1, group I, member 2); PRR; PXR; PXR.1; PXR.2; PXR1; SAR; Steroid and xenobiotic receptor; SXR
Common Name PXR
Gene Symbol NR1I2
Protein Length 111-434
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 111-434
Content And Storage Store in ultra-cold freezer (-68 to -85°C).
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Receptors
Protein Form Recombinant, Ligand Binding Domain
Form Liquid
Protein Subtype Hormone Receptors
Research Category Drug Discovery: Neurobiology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.