Learn More
Medchemexpress LLC HY-112726 5mg , PXS-4728A CAS:1478364-68-9 Purity:>98%
SDP
Catalog No. 502260555
Encompass_Preferred
HY-112726 5mg PXS-4728A CAS: 1478364-68-9 PXS-4728A (BI-1467335) is a selective, orally active inhibitor of semicarbazide-sensitive amine oxidase (SSAO). PXS-4728A ameliorates chronic obstructive pulmonary disease in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0555 Supplier Medchemexpress LLC Supplier No. HY1127265MG
HY-112726 5mg PXS-4728A CAS: 1478364-68-9 PXS-4728A (BI-1467335) is a selective, orally active inhibitor of semicarbazide-sensitive amine oxidase (SSAO). PXS-4728A ameliorates chronic obstructive pulmonary disease in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.