Learn More
Medchemexpress LLC PXS-4787 hydrochloride | 2409964-40-3 | 99.7% | 265.73 g/mol | C10H13ClFNO2S | 10 MG
SDP
Catalog No. 5000287285
Encompass_Preferred
PXS-4787 hydrochloride is a research-use small-molecule pan-lysyl oxidase (LOX) inhibitor supplied as a solid. It inhibits LOX family enzymes with low micromolar potency, reduces collagen I deposition and crosslinking in fibroblasts, and is provided with analytical documentation for laboratory studies. Not for human or veterinary use.
  • High purity (99.71%) suitable for research applications.
  • Solid form for convenient handling and storage.
  • Potent pan-LOX inhibitory activity with reported low micromolar IC50s.
  • Reduces collagen I deposition and crosslinking in cell-based assays.
  • Available in small research pack sizes for pilot and preclinical studies.
  • Provided with COA, SDS, and handling documentation.
Catalog No. 50-002-87285 Supplier Medchemexpress LLC Supplier No. HY151498A10MG
PXS-4787 hydrochloride is a research-use small-molecule pan-lysyl oxidase (LOX) inhibitor supplied as a solid. It inhibits LOX family enzymes with low micromolar potency, reduces collagen I deposition and crosslinking in fibroblasts, and is provided with analytical documentation for laboratory studies. Not for human or veterinary use.
  • High purity (99.71%) suitable for research applications.
  • Solid form for convenient handling and storage.
  • Potent pan-LOX inhibitory activity with reported low micromolar IC50s.
  • Reduces collagen I deposition and crosslinking in cell-based assays.
  • Available in small research pack sizes for pilot and preclinical studies.
  • Provided with COA, SDS, and handling documentation.