Learn More
Medchemexpress LLC PXS-4787 hydrochloride | 2409964-40-3 | C10H13ClFNO2S | 10 MM * 1 ML
SDP
Catalog No. 5000372454
Encompass_Preferred
PXS-4787 hydrochloride is a specific and effective pan-LOX (lysyl oxidase) inhibitor that abolishes lysyl oxidase activity. It inhibits LOX with IC50s of 2 μM (Bovine LOX), 3.2 μM (rh LOXL1), 0.6 μM (rh LOXL2), 1.4 μM (rh LOXL3), and 0.2 μM (rh LOXL4). This compound also reduces the deposition and crosslinking of collagen I secreted by human fibroblasts. It is for research use only and not sold to patients.
  • Specific and effective pan-LOX inhibitor
  • Abolishes lysyl oxidase activity
  • Inhibits LOX with IC50s of 2 μM (Bovine LOX), 3.2 μM (rh LOXL1), 0.6 μM (rh LOXL2), 1.4 μM (rh LOXL3), and 0.2 μM (rh LOXL4)
  • Reduces deposition and crosslinking of collagen I secreted by human fibroblasts
  • For research use only
Catalog No. 50-003-72454 Supplier Medchemexpress LLC Supplier No. HY151498A10MM/1ML
PXS-4787 hydrochloride is a specific and effective pan-LOX (lysyl oxidase) inhibitor that abolishes lysyl oxidase activity. It inhibits LOX with IC50s of 2 μM (Bovine LOX), 3.2 μM (rh LOXL1), 0.6 μM (rh LOXL2), 1.4 μM (rh LOXL3), and 0.2 μM (rh LOXL4). This compound also reduces the deposition and crosslinking of collagen I secreted by human fibroblasts. It is for research use only and not sold to patients.
  • Specific and effective pan-LOX inhibitor
  • Abolishes lysyl oxidase activity
  • Inhibits LOX with IC50s of 2 μM (Bovine LOX), 3.2 μM (rh LOXL1), 0.6 μM (rh LOXL2), 1.4 μM (rh LOXL3), and 0.2 μM (rh LOXL4)
  • Reduces deposition and crosslinking of collagen I secreted by human fibroblasts
  • For research use only