Learn More
Medchemexpress LLC PXS-5120A | 2125955-70-4 | 98.1% | 100 MG
SDP
Catalog No. 5000338588
Encompass_Preferred
PXS-5120A is a potent, irreversible fluoroallylamine inhibitor of Lysyl Oxidase-like 2/3 (LOXL2/3) with anti-fibrotic activity. It exhibits high selectivity for LOXL2 over LOXL and effectively inhibits various forms of LOXL2 and LOXL3. In vivo studies demonstrate its rapid absorption and hydrolysis to the active free base, maintaining therapeutic plasma concentrations for extended periods.
  • Potent, irreversible fluoroallylamine inhibitor of LOXL2/3
  • Exhibits anti-fibrotic activity
  • Over 300-fold selective for LOXL2 over LOXL
  • Effectively inhibits recombinant human LOXL2, human fibroblast LOXL2, recombinant mouse LOXL2, recombinant rat LOXL2, and collagen oxidation assay
  • Inhibits recombinant human LOXL3 and recombinant human LOXL4
  • Readily absorbed orally and rapidly hydrolyzed to the active free base
  • Maintains plasma concentrations above LOXL2 IC50 for prolonged periods in mice
Catalog No. 50-003-38588 Supplier Medchemexpress LLC Supplier No. HY130242100MG
PXS-5120A is a potent, irreversible fluoroallylamine inhibitor of Lysyl Oxidase-like 2/3 (LOXL2/3) with anti-fibrotic activity. It exhibits high selectivity for LOXL2 over LOXL and effectively inhibits various forms of LOXL2 and LOXL3. In vivo studies demonstrate its rapid absorption and hydrolysis to the active free base, maintaining therapeutic plasma concentrations for extended periods.
  • Potent, irreversible fluoroallylamine inhibitor of LOXL2/3
  • Exhibits anti-fibrotic activity
  • Over 300-fold selective for LOXL2 over LOXL
  • Effectively inhibits recombinant human LOXL2, human fibroblast LOXL2, recombinant mouse LOXL2, recombinant rat LOXL2, and collagen oxidation assay
  • Inhibits recombinant human LOXL3 and recombinant human LOXL4
  • Readily absorbed orally and rapidly hydrolyzed to the active free base
  • Maintains plasma concentrations above LOXL2 IC50 for prolonged periods in mice