Learn More
Medchemexpress LLC PXS-5120A | 2125955-70-4 | 98.1% | C22H25ClFN3O4S | 10MG
SDP
Catalog No. 5000261503
Encompass_Preferred
PXS-5120A is a potent, irreversible fluoroallylamine inhibitor of lysyl oxidase-like enzymes LOXL2 and LOXL3, described in the medicinal chemistry literature as a mechanism-based inhibitor with anti-fibrotic activity and high selectivity for LOXL2. It is supplied for research use and has been characterized by CAS, formula, and molecular weight in supplier and public databases.
  • Irreversible mechanism-based inhibitor of LOXL2 and LOXL3.
  • Greater than 300-fold selectivity for LOXL2 versus LOX.
  • Reported Ki ~83 nM for LOXL2 and pIC50 ≈ 8.4.
  • Suitable for anti-fibrotic and LOXL biology research.
  • Characterized molecular formula and molecular weight for analytical use.
Catalog No. 50-002-61503 Supplier Medchemexpress LLC Supplier No. HY13024210MG
PXS-5120A is a potent, irreversible fluoroallylamine inhibitor of lysyl oxidase-like enzymes LOXL2 and LOXL3, described in the medicinal chemistry literature as a mechanism-based inhibitor with anti-fibrotic activity and high selectivity for LOXL2. It is supplied for research use and has been characterized by CAS, formula, and molecular weight in supplier and public databases.
  • Irreversible mechanism-based inhibitor of LOXL2 and LOXL3.
  • Greater than 300-fold selectivity for LOXL2 versus LOX.
  • Reported Ki ~83 nM for LOXL2 and pIC50 ≈ 8.4.
  • Suitable for anti-fibrotic and LOXL biology research.
  • Characterized molecular formula and molecular weight for analytical use.