Learn More
Medchemexpress LLC PXS-5120A | 2125955-70-4 | MFCD32263436 | 98.1% | 481.97 | C22H25ClFN3O4S | 5 MG
SDP
Catalog No. 5000262363
Encompass_Preferred
PXS-5120A is a research-grade, irreversible fluoroallylamine inhibitor selective for lysyl oxidase-like 2 and 3 (LOXL2/3) with reported anti-fibrotic activity. It demonstrates high potency for LOXL2 (Ki = 83 nM; pIC50 = 8.4) and >300-fold selectivity over LOXL, and is supplied for laboratory research use.
  • Potent irreversible LOXL2/3 inhibitor.
  • High selectivity for LOXL2 (Ki = 83 nM).
  • Reported anti-fibrotic activity suitable for pathway studies.
  • Supplied with reported purity of 98.12%.
  • Available in milligram quantities and as a DMSO solution.
Catalog No. 50-002-62363 Supplier Medchemexpress LLC Supplier No. HY1302425MG
PXS-5120A is a research-grade, irreversible fluoroallylamine inhibitor selective for lysyl oxidase-like 2 and 3 (LOXL2/3) with reported anti-fibrotic activity. It demonstrates high potency for LOXL2 (Ki = 83 nM; pIC50 = 8.4) and >300-fold selectivity over LOXL, and is supplied for laboratory research use.
  • Potent irreversible LOXL2/3 inhibitor.
  • High selectivity for LOXL2 (Ki = 83 nM).
  • Reported anti-fibrotic activity suitable for pathway studies.
  • Supplied with reported purity of 98.12%.
  • Available in milligram quantities and as a DMSO solution.