Learn More
Medchemexpress LLC HY-114286A 5mg Medchemexpress, PXS-5153A (monohydrochloride) CAS: Purity:>98%
SDP
Catalog No. 501969321
Encompass_Preferred
Medchemexpress, HY-114286A 5mg PXS-5153A (monoHCl) CAS: PXS-5153A monohydrochloride is a potent, selective, orally active and fast-acting lysyl oxidase like 2/3 enzymatic (LOXL2/LOXL3) inhibitor, with an IC50 of <40 nM for LOXL2 across all mammalian species and an IC50 of 63 nM for human LOXL3. PXS-5153A monohydrochloride could reduce crosslinks and ameliorates fibrosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9321 Supplier Medchemexpress LLC Supplier No. HY114286A5MG
Medchemexpress, HY-114286A 5mg PXS-5153A (monoHCl) CAS: PXS-5153A monohydrochloride is a potent, selective, orally active and fast-acting lysyl oxidase like 2/3 enzymatic (LOXL2/LOXL3) inhibitor, with an IC50 of <40 nM for LOXL2 across all mammalian species and an IC50 of 63 nM for human LOXL3. PXS-5153A monohydrochloride could reduce crosslinks and ameliorates fibrosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.