Learn More
Medchemexpress LLC PXS-5153A (monohydrochloride) | 2125956-82-1 | 99.6% | 438.95 | 50 MG
SDP
Catalog No. 5000437570
Encompass_Preferred
PXS-5153A monohydrochloride is a potent, selective, orally active and fast-acting lysyl oxidase like 2/3 enzymatic (LOXL2/LOXL3) inhibitor, with an IC50 of <40 nM for LOXL2 across all mammalian species and an IC50 of 63 nM for human LOXL3. PXS-5153A monohydrochloride could reduce crosslinks and ameliorate fibrosis.
  • Exhibits an IC50 of <40 nM for LOXL2 across all mammalian species tested.
  • Also inhibits human LOXL3 with an IC50 value of 63 nM.
  • Is >40-fold selective for LOXL2 over both LOX and LOXL1 and >700-fold selective over other related amine oxidases.
  • Is a fast acting inhibitor, with enzymatic activity almost entirely blocked within 15 minutes.
  • Reduces crosslinks and ameliorates fibrosis.
Catalog No. 50-004-37570 Supplier Medchemexpress LLC Supplier No. HY114286A50MG
PXS-5153A monohydrochloride is a potent, selective, orally active and fast-acting lysyl oxidase like 2/3 enzymatic (LOXL2/LOXL3) inhibitor, with an IC50 of <40 nM for LOXL2 across all mammalian species and an IC50 of 63 nM for human LOXL3. PXS-5153A monohydrochloride could reduce crosslinks and ameliorate fibrosis.
  • Exhibits an IC50 of <40 nM for LOXL2 across all mammalian species tested.
  • Also inhibits human LOXL3 with an IC50 value of 63 nM.
  • Is >40-fold selective for LOXL2 over both LOX and LOXL1 and >700-fold selective over other related amine oxidases.
  • Is a fast acting inhibitor, with enzymatic activity almost entirely blocked within 15 minutes.
  • Reduces crosslinks and ameliorates fibrosis.