Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PXS-5505 anhydrous (LOX-IN-3 dihydrochloride) | 2409964-23-2 | 99.9% | 353.24 g·mol⁻¹ | C13H15Cl2FN2O2S | 5 MG
SDP

Catalog No. 5000221436
Encompass_Preferred

PXS-5505 anhydrous (LOX-IN-3 dihydrochloride) is an orally active pan-lysyl oxidase inhibitor supplied as an anhydrous dihydrochloride solid for preclinical research. It inhibits LOX/LOXL family enzymes and is commonly used to investigate fibrosis, cancer, and angiogenesis. The material is provided at high purity and includes recommended storage guidance to maintain stability.

  • Orally active pan-lysyl oxidase inhibitor for LOX/LOXL studies.
  • Suitable for fibrosis, cancer, and angiogenesis research.
  • Anhydrous dihydrochloride solid form for reproducible handling.
  • High purity appropriate for in vitro and in vivo experiments.
  • Recommended sealed storage away from moisture to preserve stability.
  • Molecular weight 353.24 g·mol⁻¹; formula C13H15Cl2FN2O2S.

Catalog No. 50-002-21436 Supplier Medchemexpress LLC Supplier No. HY138625A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PXS-5505 anhydrous (LOX-IN-3 dihydrochloride) is an orally active pan-lysyl oxidase inhibitor supplied as an anhydrous dihydrochloride solid for preclinical research. It inhibits LOX/LOXL family enzymes and is commonly used to investigate fibrosis, cancer, and angiogenesis. The material is provided at high purity and includes recommended storage guidance to maintain stability.

  • Orally active pan-lysyl oxidase inhibitor for LOX/LOXL studies.
  • Suitable for fibrosis, cancer, and angiogenesis research.
  • Anhydrous dihydrochloride solid form for reproducible handling.
  • High purity appropriate for in vitro and in vivo experiments.
  • Recommended sealed storage away from moisture to preserve stability.
  • Molecular weight 353.24 g·mol⁻¹; formula C13H15Cl2FN2O2S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.