Learn More
Medchemexpress LLC Pxs-5505 anhydrous (LOX-IN-3 dihydrochloride) | 2409964-23-2 | 99.9% | C13H15Cl2FN2O2S | 10MG
SDP
Catalog No. 5000261282
Encompass_Preferred
PXS-5505 anhydrous (LOX-IN-3 dihydrochloride) is an orally active pan-lysyl oxidase (LOX) inhibitor supplied as an anhydrous dihydrochloride salt for laboratory research. It is used in preclinical studies of fibrosis, cancer, and angiogenesis, and is provided at high reported purity for experimental applications (for research use only).
  • Inhibits lysyl oxidase family enzymes for fibrosis and tumor microenvironment studies.
  • Anhydrous dihydrochloride salt form suitable for storage and handling.
  • High reported purity (99.9%) for reproducible experimental results.
  • Supplied in small-scale packaging appropriate for preclinical research.
  • Datasheet and safety data sheet available to support handling and protocols.
Catalog No. 50-002-61282 Supplier Medchemexpress LLC Supplier No. HY138625A10MG
PXS-5505 anhydrous (LOX-IN-3 dihydrochloride) is an orally active pan-lysyl oxidase (LOX) inhibitor supplied as an anhydrous dihydrochloride salt for laboratory research. It is used in preclinical studies of fibrosis, cancer, and angiogenesis, and is provided at high reported purity for experimental applications (for research use only).
  • Inhibits lysyl oxidase family enzymes for fibrosis and tumor microenvironment studies.
  • Anhydrous dihydrochloride salt form suitable for storage and handling.
  • High reported purity (99.9%) for reproducible experimental results.
  • Supplied in small-scale packaging appropriate for preclinical research.
  • Datasheet and safety data sheet available to support handling and protocols.