Learn More
TARGETMOL CHEMICALS INC PXS-6302 hydrochloride 1MG
SDP
Catalog No. 502876092
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. PXS-6302 hydrochloride is a potent irreversible lysine oxidase (LOX) inhibitor, inhibiting Bovine LOX, rh LOXL1, rh LOXL2, rh LOXL3, and rh LOXL4 with IC50 of 3.7 uM, 3.4 uM, 0.4 uM, 1.5 uM, and 0.3 uM, respectively. PXS-6302 hydrochloride penetrates easily through the skin and is able to reduce collagen accumulation, significantly improving the appearance of scars. Purity 99.89%
Catalog No. 50-287-6092 Supplier TARGETMOL CHEMICALS INC Supplier No. T60438L1MG
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. PXS-6302 hydrochloride is a potent irreversible lysine oxidase (LOX) inhibitor, inhibiting Bovine LOX, rh LOXL1, rh LOXL2, rh LOXL3, and rh LOXL4 with IC50 of 3.7 uM, 3.4 uM, 0.4 uM, 1.5 uM, and 0.3 uM, respectively. PXS-6302 hydrochloride penetrates easily through the skin and is able to reduce collagen accumulation, significantly improving the appearance of scars. Purity 99.89%