Learn More
Medchemexpress LLC PXS-6302 hydrochloride | 2584947-79-3 | C10H11ClF3NO2S | 1 MG
SDP
Catalog No. 5000372205
Encompass_Preferred
PXS-6302 hydrochloride is an irreversible lysyl oxidase inhibitor. It demonstrates high permeability across cell monolayers and reduces collagen deposition. This compound has shown significant improvement in scar appearance in porcine models of excisional and burn injury under topical application.
  • Irreversible lysyl oxidase inhibitor
  • Reduces collagen deposition
  • Significantly improves scar appearance
  • Readily skin penetrability
Catalog No. 50-003-72205 Supplier Medchemexpress LLC Supplier No. HY151499A1MG
PXS-6302 hydrochloride is an irreversible lysyl oxidase inhibitor. It demonstrates high permeability across cell monolayers and reduces collagen deposition. This compound has shown significant improvement in scar appearance in porcine models of excisional and burn injury under topical application.
  • Irreversible lysyl oxidase inhibitor
  • Reduces collagen deposition
  • Significantly improves scar appearance
  • Readily skin penetrability