Learn More
Medchemexpress LLC PXS-6302 hydrochloride | 2584947-79-3 | C10H11ClF3NO2S | 5 MG
SDP
Catalog No. 5000372561
Encompass_Preferred
PXS-6302 hydrochloride is an irreversible lysyl oxidase inhibitor that effectively targets multiple LOX enzymes, with IC50 values ranging from 0.3 μM to 3.7 μM. Its excellent skin penetrability facilitates the reduction of collagen deposition and significantly enhances the appearance of scars.
  • Acts as an irreversible lysyl oxidase inhibitor
  • Effective against various LOX enzyme types
  • Exhibits excellent skin penetrability
  • Reduces collagen deposition
  • Significantly improves scar appearance
  • High permeability observed in Caco-2 and MDCKII cell assays
  • Inhibits LOX and reduces crosslinking in porcine injury models
  • Topical application improves scar appearance in murine and porcine models
Catalog No. 50-003-72561 Supplier Medchemexpress LLC Supplier No. HY151499A5MG
PXS-6302 hydrochloride is an irreversible lysyl oxidase inhibitor that effectively targets multiple LOX enzymes, with IC50 values ranging from 0.3 μM to 3.7 μM. Its excellent skin penetrability facilitates the reduction of collagen deposition and significantly enhances the appearance of scars.
  • Acts as an irreversible lysyl oxidase inhibitor
  • Effective against various LOX enzyme types
  • Exhibits excellent skin penetrability
  • Reduces collagen deposition
  • Significantly improves scar appearance
  • High permeability observed in Caco-2 and MDCKII cell assays
  • Inhibits LOX and reduces crosslinking in porcine injury models
  • Topical application improves scar appearance in murine and porcine models