Learn More
Medchemexpress LLC PXS-6302 hydrochloride | 2584947-79-3 | MFCD09749328 | 99.8% | 301.71 | C10H11ClF3NO2S | 10 MG
SDP
Catalog No. 5000287417
Encompass_Preferred
PXS-6302 hydrochloride is a small-molecule, irreversible pan-lysyl oxidase (LOX) inhibitor used in research to reduce collagen crosslinking and improve scar appearance. It has been evaluated in topical formulations and preclinical models and is used for in vitro and topical studies of fibrosis, scar formation, and extracellular matrix remodeling.
  • Reduces collagen deposition and crosslinking in skin models.
  • Suitable for topical formulation research due to skin penetrability.
  • Irreversible inhibitor of LOX family with low-micromolar potency.
  • Available in research-grade purity (>99% by HPLC).
  • Stable under sealed storage; follow recommended cold storage conditions.
Catalog No. 50-002-87417 Supplier Medchemexpress LLC Supplier No. HY151499A10MG
PXS-6302 hydrochloride is a small-molecule, irreversible pan-lysyl oxidase (LOX) inhibitor used in research to reduce collagen crosslinking and improve scar appearance. It has been evaluated in topical formulations and preclinical models and is used for in vitro and topical studies of fibrosis, scar formation, and extracellular matrix remodeling.
  • Reduces collagen deposition and crosslinking in skin models.
  • Suitable for topical formulation research due to skin penetrability.
  • Irreversible inhibitor of LOX family with low-micromolar potency.
  • Available in research-grade purity (>99% by HPLC).
  • Stable under sealed storage; follow recommended cold storage conditions.