Learn More
Medchemexpress LLC Pxs-6302 hydrochloride | 2584947-79-3 | MFCD09749328 | C10H11ClF3NO2S | 25 MG
SDP
Catalog No. 5000374518
Encompass_Preferred
PXS-6302 hydrochloride is an irreversible lysyl oxidase inhibitor with IC50s of 3.7 μM (Bovine LOX), 3.4 μM (rh LOXL1), 0.4 μM (rh LOXL2), 1.5 μM (rh LOXL3), and 0.3 μM (rh LOXL4). It exhibits good skin penetrability, reduces collagen deposition, and significantly improves scar appearance.
  • Irreversible lysyl oxidase inhibitor
  • Exhibits good skin penetrability
  • Reduces collagen deposition
  • Significantly improves scar appearance
  • Reduces crosslinking
Catalog No. 50-003-74518 Supplier Medchemexpress LLC Supplier No. 25MGHY151499A
PXS-6302 hydrochloride is an irreversible lysyl oxidase inhibitor with IC50s of 3.7 μM (Bovine LOX), 3.4 μM (rh LOXL1), 0.4 μM (rh LOXL2), 1.5 μM (rh LOXL3), and 0.3 μM (rh LOXL4). It exhibits good skin penetrability, reduces collagen deposition, and significantly improves scar appearance.
  • Irreversible lysyl oxidase inhibitor
  • Exhibits good skin penetrability
  • Reduces collagen deposition
  • Significantly improves scar appearance
  • Reduces crosslinking