Learn More
Medchemexpress LLC HY-141644 10mg , PY-60 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14164410MG
Encompass_Preferred
HY-141644 10mg PY-60 CAS: PY-60 is a robust and specific activator of YAP transcriptional activity that targets annexin A2 (ANXA2) with a Kd of 1.4 µM. PY-60 directly binds to ANXA2 and antagonizes its normal cellular function of repressing YAP activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0125