Learn More
Medchemexpress LLC PY-60 | 2765218-56-0 | 99.4% | C16H15N3O2S | 3.1 MG
SDP
Catalog No. 5000353906
Encompass_Preferred
PY-60 is a potent and specific activator of YAP transcriptional activity, targeting annexin A2 (ANXA2) with a Kd of 1.4 μM. It directly binds to ANXA2, counteracting its normal function of repressing YAP activity. This compound dose-dependently induces luciferase activity in 293A-TEAD-LUC cells and promotes nuclear localization of YAP, while significantly increasing YAP-controlled transcripts in human cell lines.
  • Targets annexin A2 to activate YAP
  • Induces luciferase activity in 293A-TEAD-LUC cells
  • Promotes YAP and TEAD protein association
  • Increases YAP nuclear localization at high cell density
  • Elevates YAP-controlled transcripts (ANRKD1, CYR61, CTGF)
  • Liberates ANXA2-YAP complex from cell membrane
  • Activates proproliferative, YAP-dependent program
  • Promotes keratinocyte expansion and epidermal thickening in vivo
Catalog No. 50-003-53906 Supplier Medchemexpress LLC Supplier No. HY14164410MM/1ML
PY-60 is a potent and specific activator of YAP transcriptional activity, targeting annexin A2 (ANXA2) with a Kd of 1.4 μM. It directly binds to ANXA2, counteracting its normal function of repressing YAP activity. This compound dose-dependently induces luciferase activity in 293A-TEAD-LUC cells and promotes nuclear localization of YAP, while significantly increasing YAP-controlled transcripts in human cell lines.
  • Targets annexin A2 to activate YAP
  • Induces luciferase activity in 293A-TEAD-LUC cells
  • Promotes YAP and TEAD protein association
  • Increases YAP nuclear localization at high cell density
  • Elevates YAP-controlled transcripts (ANRKD1, CYR61, CTGF)
  • Liberates ANXA2-YAP complex from cell membrane
  • Activates proproliferative, YAP-dependent program
  • Promotes keratinocyte expansion and epidermal thickening in vivo