Learn More
Medchemexpress LLC Py-maa-val-cit-pab-mmae | 2247398-68-9 | 98.5% | 1446.79 | C72H111N13O16S | 50 MG
SDP
Catalog No. 5000202298
Encompass_Preferred
Py-MAA-Val-Cit-PAB-MMAE is a drug-linker conjugate used as a building block in antibody-drug conjugate (ADC) synthesis. It links the cytotoxic payload MMAE to a protease-cleavable Val-Cit-PAB linker via a Py-MAA spacer and is intended for research use in ADC development.
  • Protease-cleavable Val-Cit-PAB linker for controlled payload release
  • Contains a Py-MAA spacer for conjugation flexibility
  • Includes monomethyl auristatin E (MMAE) as the cytotoxic payload
  • Supplied as a dry material suitable for organic solvent handling
  • Intended for research and development applications in ADC synthesis
Catalog No. 50-002-02298 Supplier Medchemexpress LLC Supplier No. HY14887550MG
Py-MAA-Val-Cit-PAB-MMAE is a drug-linker conjugate used as a building block in antibody-drug conjugate (ADC) synthesis. It links the cytotoxic payload MMAE to a protease-cleavable Val-Cit-PAB linker via a Py-MAA spacer and is intended for research use in ADC development.
  • Protease-cleavable Val-Cit-PAB linker for controlled payload release
  • Contains a Py-MAA spacer for conjugation flexibility
  • Includes monomethyl auristatin E (MMAE) as the cytotoxic payload
  • Supplied as a dry material suitable for organic solvent handling
  • Intended for research and development applications in ADC synthesis