Learn More
Biolog Inc PY XYLAN - Peptone Yeast Extract Broth with Xylan, 10 Tubes/Pkg
SDP
Catalog No. NC1424891
Encompass
This item has a minimum qty of 15 per supplier requirements.
Peptone Yeast Extract Broth Based Media are enriched nonselective broth used for growth & identification of anaerobic bacteria based on their biochemical features. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.PY Broth contains hemin & vitamin K for the recovery of fastidious organisms such as Prevotella spp and Porphyromonas spp. PRAS (Pre-reduced anaerobically sterilized) biochemicals are considered the gold standard for biochemical testing and characterization of anaerobes. Adding specific biochemical, the Peptone Yeast Extract Broth Based Media can determine whether anaerobic bacteria will utilize the biochemical or if growth is stimulated or inhibited.
This item is not returnable. View return policy
Catalog No. NC1424891 Supplier Biolog Inc Supplier No. AS859
This item has a minimum qty of 15 per supplier requirements.
Peptone Yeast Extract Broth Based Media are enriched nonselective broth used for growth & identification of anaerobic bacteria based on their biochemical features. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.PY Broth contains hemin & vitamin K for the recovery of fastidious organisms such as Prevotella spp and Porphyromonas spp. PRAS (Pre-reduced anaerobically sterilized) biochemicals are considered the gold standard for biochemical testing and characterization of anaerobes. Adding specific biochemical, the Peptone Yeast Extract Broth Based Media can determine whether anaerobic bacteria will utilize the biochemical or if growth is stimulated or inhibited.