Learn More
Enzo Life Sciences [pTyr1146/1150] Insulin receptor (1142-1153) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP3210500
Encompass_Preferred
This sequence, TRDIpYETDpYYRK, is from the insulin receptor ß-subunit cytoplasmic domain, inclusive of regulatory autophosphorylation sites Tyr1146, Tyr1150 and Tyr1151. The unphosphorylated version of this peptide has been used as an insulin receptor kinase substrate. The phosphorylated peptide has been used as a protein tyrosine phosphatase substrate. Long Term Storage: -20°C.
Catalog No. 50-201-1345