Learn More
Medchemexpress LLC HY-23926 5mg Medchemexpress, PYBG CAS:680622-71-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY239265MG
Encompass_Preferred
Medchemexpress, HY-23926 5mg PYBG CAS:680622-71-3 PYBG acts as a versatile precursor to be facilely conjugated with various fluorescent dyes through ‘Click chemistry’ and Sonogashira coupling reactions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2222