Learn More
Apexbio Technology LLC PyBOP, 128625-52-5, MFCD00077411, 100 g
SDP
Catalog No. 501150898
Encompass_Preferred
MF: C18H28F6N6OP2, MW: 520.4, Solubility: 52mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. PyBOP (benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate), the analog of BOP where dimethylamino groups are replaced with pyrrolidino, is a peptide coupling reagent used in solid phase peptide synthesis.
Catalog No. 50-115-0898 Supplier Apexbio Technology LLC Supplier No. A7029100000
MF: C18H28F6N6OP2, MW: 520.4, Solubility: 52mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. PyBOP (benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate), the analog of BOP where dimethylamino groups are replaced with pyrrolidino, is a peptide coupling reagent used in solid phase peptide synthesis.