Learn More
Aviva Systems Biology Corp PYCARD Antibody - C-terminal region (ARP36561_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36561_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PYCARD. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PYCARD. Alternate names: ASC, CARD5, MGC10332, TMS, TMS-1, TMS1ApplicationWB. Species reactivity: Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-284-501