Learn More
Aviva Systems Biology Corp PYCR1 Antibody - middle region (ARP57751_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57751_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PYCR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PYCR1. Alternate names: P5C, P5CR, PIG45, PP222, PYCR, PRO3, ARCL2B, ARCL3BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62143