Learn More
Aviva Systems Biology Corp PYCR2 Antibody - C-terminal region (ARP54938_P050) | 100uL
SDP
Catalog No. 5001461373
Encompass_Preferred
This is a rabbit polyclonal antibody against PYCR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PYCR2. Alternate names: P5CR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61373 Supplier Aviva Systems Biology Corp Supplier No. ARP54938_P050
This is a rabbit polyclonal antibody against PYCR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PYCR2. Alternate names: P5CR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat