Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyd-106 | 1560894-05-4 | 98.1% | 432.47 | C25H24N2O5 | 50 MG
SDP

Catalog No. 5000228781
Encompass_Preferred

A stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors. Supplied as an off-white to light-yellow solid in a 50 mg package, this compound is intended for research use only and is accompanied by a product data sheet and safety data sheet.

  • Stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors.
  • High purity (98.13%) suitable for biochemical and electrophysiological studies.
  • Chemical formula C25H24N2O5 and molecular weight 432.47 g/mol.
  • Appearance: off-white to light-yellow solid; packaged as 50 mg aliquots.
  • Recommended storage: powder at -20°C for up to 3 years; in solvent at -80°C for long-term storage.
  • Includes downloadable data sheet and SDS for laboratory safety and handling.

Catalog No. 50-002-28781 Supplier Medchemexpress LLC Supplier No. HY11773450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors. Supplied as an off-white to light-yellow solid in a 50 mg package, this compound is intended for research use only and is accompanied by a product data sheet and safety data sheet.

  • Stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors.
  • High purity (98.13%) suitable for biochemical and electrophysiological studies.
  • Chemical formula C25H24N2O5 and molecular weight 432.47 g/mol.
  • Appearance: off-white to light-yellow solid; packaged as 50 mg aliquots.
  • Recommended storage: powder at -20°C for up to 3 years; in solvent at -80°C for long-term storage.
  • Includes downloadable data sheet and SDS for laboratory safety and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.