Learn More
Medchemexpress LLC PYD-106 | 1560894-05-4 | 98.1% | 432.47 g/mol | C25H24N2O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11773410MG
Encompass_Preferred
PYD-106 is a stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors that increases channel opening frequency and potentiates receptor function. It is intended for research use.
  • Selective positive allosteric modulator of GluN2C-containing NMDA receptors.
  • Increases channel opening frequency and potentiates receptor activity.
  • High purity: 98.13%.
  • Chemical formula C25H24N2O5; molecular weight 432.47 g/mol.
  • Provided in 10 mg packaging for laboratory research.
Catalog No. 50-002-26565