Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyd-106 | 1560894-05-4 | MFCD06797172 | 98.1% | 432.47 g/mol | C25H24N2O5 | 25 MG
SDP

Catalog No. 5000228224
Encompass_Preferred

PYD-106 is a stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors. It potentiates channel opening frequency and duration and is provided as a research-grade small-molecule tool for pharmacology studies.

  • Selective positive allosteric modulator of GluN2C-containing NMDA receptors.
  • Increases channel open frequency and open duration at effective concentrations.
  • High purity (98.13%) suitable for research applications.
  • Molecular weight 432.47 g/mol; formula C25H24N2O5.
  • Recommended storage: powder at -20°C for long-term stability.
  • Available in a 25 mg research pack.

Catalog No. 50-002-28224 Supplier Medchemexpress LLC Supplier No. HY11773425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PYD-106 is a stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors. It potentiates channel opening frequency and duration and is provided as a research-grade small-molecule tool for pharmacology studies.

  • Selective positive allosteric modulator of GluN2C-containing NMDA receptors.
  • Increases channel open frequency and open duration at effective concentrations.
  • High purity (98.13%) suitable for research applications.
  • Molecular weight 432.47 g/mol; formula C25H24N2O5.
  • Recommended storage: powder at -20°C for long-term stability.
  • Available in a 25 mg research pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.