Learn More
MOLTOX Molecular Toxicology Inc Phytone Yeast Extract (PYE) Agar, 500 ml/bottle, Store in Refrigerator
SDP
Catalog No. 50948834
Encompass_Preferred
Phytone Yeast Extract Agar, or PYE, is used as a selective isolation medium of dermatophytes and other pathogenic fungi. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-834 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S072
Phytone Yeast Extract Agar, or PYE, is used as a selective isolation medium of dermatophytes and other pathogenic fungi. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.