Learn More
FUJIFILM Biosciences Pyflubumide Metabolite B Stand
Catalog No. 5000450431
Encompass_Preferred
Reagent for Pesticide Residue Analysis Maximum residue limits for pyflubumide are the sum of residues of pyflubumide and 3-isobutyl-135-trimethyl-4- 222-trifluoro-1-methoxy-1-trifluoromethylethyl pyrazole-4-carboxanilide metabolite B calculated as mepanipyrim White to slightly yellowish brown crystalline powder Used for residue testing in environmental and agricultural samples
Catalog No. 50-004-50431 Supplier FUJIFILM Biosciences Supplier No. 16027551
Reagent for Pesticide Residue Analysis Maximum residue limits for pyflubumide are the sum of residues of pyflubumide and 3-isobutyl-135-trimethyl-4- 222-trifluoro-1-methoxy-1-trifluoromethylethyl pyrazole-4-carboxanilide metabolite B calculated as mepanipyrim White to slightly yellowish brown crystalline powder Used for residue testing in environmental and agricultural samples