Learn More
Biolog Inc Peptone Yeast Extract Broth with Glucose & Gelatin (PYG GELATIN), 10/pk
SDP
Catalog No. NC0995281
Encompass
This item has a minimum qty of 15 per supplier requirements.
Peptone Yeast Extract Broth Based Media are enriched nonselective media that are supplemented with hemin and vitamin K1 to facilitate the recovery of the more fastidious organisms such as Prevotella spp, Porphyromonas spp, and the Bacteroides fragilis group. Resazurin is added as a redox indicator.
Catalog No. NC0995281 Supplier Biolog Inc Supplier No. AS860
This item has a minimum qty of 15 per supplier requirements.
Peptone Yeast Extract Broth Based Media are enriched nonselective media that are supplemented with hemin and vitamin K1 to facilitate the recovery of the more fastidious organisms such as Prevotella spp, Porphyromonas spp, and the Bacteroides fragilis group. Resazurin is added as a redox indicator.