Learn More
Aviva Systems Biology Corp Pygm Antibody - C-terminal region (ARP48678_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48678_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pygm. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI115133, PGApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-375