Learn More
Aviva Systems Biology Corp PYGO2 Antibody - N-terminal region (ARP51238_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51238_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PYGO2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PYGO2. Alternate names: 1190004M21Rik, FLJ33226ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60525