Learn More
Sino Biological Recombinant Human AMPK (A2/B1/G2) Heterotrimer Protein (Full Length, A2/B1/G2, His & GST Tag), Active
Catalog No. 502573520
Encompass_Preferred
Recombinant full length human AMPK (combination of A2/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using N-terminal GST and C-terminal His tags.
Catalog No. 50-257-3520 Supplier Sino Biological Supplier No. P8410GH500
Recombinant full length human AMPK (combination of A2/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using N-terminal GST and C-terminal His tags.