Learn More
Serim Research Corp Serim PyloriTek Helicobacter pylori Test Kit
SDP
Catalog No. NC1078535
Encompass_Preferred
Test detects urease activity in gastric biopsy specimens used as a presumptive identification of Helicobacter pylori infections. Results available within 1 hour. Kit contains 20 tests
Catalog No. NC1078535 Supplier Serim Research Corp Supplier No. 5140K
Test detects urease activity in gastric biopsy specimens used as a presumptive identification of Helicobacter pylori infections. Results available within 1 hour. Kit contains 20 tests